Learn More
DWK Life Sciences Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Product Code. p-7189349 Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 32
GL 45
GL 54
GL 60
Unit Size:
Pack of 10
Product Code. 11728011
Brand: DWK Life Sciences 293031901
Only null left
Add to basket
Edge
Add to basket
Tamper evident cap for wide mouth/short form DIN soda lime glass bottle thread.
Tamper evident, blue polypropylene, linerless screw cap for wide mouth/short form DIN thread soda lime glass bottles.
  • Tamper evident ring
  • LInerless
  • Short form DIN thread
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Storage of samples and reagents; Sample collection; Transport and shipping
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GL 32
Diameter (Metric) 44 mm
Autoclavable Autoclavable