missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tamsulosin hydrochloride
GREENER_CHOICE

Product Code. 18772764
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772764 10 mg 10mg
18762754 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18772764 Supplier Tocris Bioscience™ Supplier No. 3050/10
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Selective α1A antagonist

Tamsulosin hydrochloride is a selective α1A-adrenoceptor antagonist (pKi values are 9.97, 9.64 and 8.86 for α1A, α1B and α1D subtypes respectively). Decreases resting maximal urethral pressure with negligible effect on mean arterial blood pressure in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106463-17-6
Molecular Formula C20H29ClN2O5S

Specifications

Quantity 10 mg
Formula Weight 444.97
Purity >99%
Molecular Weight (g/mol) 444.971
Chemical Name or Material Tamsulosin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.