missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ EDG3-bla U2OS Cells

Product Code. 10165694
Click to view available options
Quantity:
8 x 106 cells/vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10165694

Brand: Thermo Scientific™ K1848

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tango™ EDG3-bla U2OS cells contain the human Endothelial Differentiation Gene 3 (EDG3) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

Tango™ EDG3-bla U2OS cells contain the human Endothelial Differentiation Gene 3 (EDG3) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ EDG3-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of S1P. In addition, Tango™ EDG3-bla U2OS cells have been tested for assay performance under variable conditions.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005226.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage Tango™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Cytoplasmic Tail Native Tail
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Pool of Stable Cells
Product Line Tango
Quantity 8 x 106 cells/vial
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDG3
Type Dividing Cells
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt