Få mere at vide
Thermo Scientific™ Tango™ NPY5R-bla U2OS Cells
Beskrivelse
The Tango™ NPY5R -bla U2OS cells contain the human Neuropeptide Y Receptor Y5 (NPY5R) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ NPY5R-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ NPY5R-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Tekniske data
Tekniske data
| Accession Number | NM_006174.2 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ NPY5R-bla U2OS cells • Tango™ NPY5R-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ NPY5R-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Cytoplasmic Tail | Tail from AVPR2 GPCR |
| Description | Tango™ NPY5R-bla U2OS Cells, 8 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays |
| Gene Symbol | NPY5R |
| Includes | Tango™ NPY5R-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ NPY5R-bla U2OS cells, Tango™ NPY5R-bla U2OS assay protocol, certificate of analysis |
| Vis mere |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.