missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid, 95%

Product Code. 10174070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174070 500 g Plastic bottle 500g
10702631 5 g Glass bottle 5g
10702441 1 kg Plastic bottle 1kg
10484745 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10174070 Supplier Thermo Scientific Acros Supplier No. 202425000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Melting Point 218.0°C
Color White to Yellow
Density 198g/mL
Quantity 500 g
Flash Point 198°C
Infrared Spectrum Authentic
Loss on Drying 12.0% max.
Packaging Plastic bottle
Physical Form Beads
Assay Percent Range 95%
Merck Index 15, 9184
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: soluble in glycerol, ethanol and acetone;, practically insoluble in benzene, chcl3, diethyl e, ther, petroleum ether, cs2, ccl4, hexane;, slightly soluble in anhydrous ethanol.
Formula Weight 1701.23
Percent Purity 95%
Chemical Name or Material Tannic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.