missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tannic acid, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 218.0°C |
| Color | White to Yellow |
| Assay Percent Range | 95% |
| Merck Index | 15, 9184 |
| Solubility Information | Solubility in water: 250g/L (20°C). Other solubilities: soluble in glycerol, ethanol and acetone;, practically insoluble in benzene, chcl3, diethyl e, ther, petroleum ether, cs2, ccl4, hexane;, slightly soluble in anhydrous ethanol. |
| Formula Weight | 1701.23 |
| Percent Purity | 95% |
| Density | 198g/mL |
| Quantity | 5 g |
| Flash Point | 198°C |
| Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur