missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid, ACS

Product Code. p-3208647
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11307878

Brand: Thermo Scientific Alfa Aesar 036410.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol

Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Melting Point ∽218°C (decomposition)
Beilstein 8186396
Solubility Information Soluble in water,warm glycerol,alcohol and acetone.
Formula Weight 1701.23
Grade ACS Reagent
Quantity 100 g
Flash Point 199°C (390°F)
Physical Form Powder
Sensitivity Air and light sensitive
Chemical Name or Material Tannic Acid
compliance-icons
Product Identifier
  • Tannic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tannic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.