missing translation for 'onlineSavingsMsg'
Learn More

Tannic Acid, TRC

Product Code. 30417019
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417019 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417019 Supplier Toronto Research Chemicals Supplier No. TRCT006560100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tannin (Tannic acid)
CAS 1401-55-4
Molecular Formula C76H52O46
Quantity 100 g
Synonym Tannic Acids, Tannin analysis, AL, AL (tannin), Arbor Lock 20, BM 539, Bondtite 241, Bondtite 345, Bondtite 645, Brewtan, Brewtan C, Colatan GT 100, Crown ATO, DT-R 240, Excellence Vintage, F-Tannin, FS 500M, Fintan OP, Floctan 1, Floctan 3, Fresh Shiraimatsu FS 500M, Gallotannic Acid, Gallotannic Acids, Gallotannin, Gallotannins, Hifix 45, Hifix SL, Hifix SLA, Hifix SW-A, Kakitafu, Lexsod P, MP-TR, Makubunri Seisei Kaki Tannin H 1, Pancil, Pancil FG 22, Pancil FG 30, Pancil FG 70, Quertanil, Resorcinex Pecan Tannin 9901L, SV 82, Sunlife TN, TW 75, Tanal 02C, Tanal 1, Tanaphen P 500, Tanex A-FCC, Tanex RS 93, Tannic Acid KT, Tannic Acid ME, Tannic Acid X, Tannic acid, Tannic acid AL, Tannin, Textan 3, UT 10, UT 10 (tannin), Vitanil B, Vitanil IM, Weibull
InChI Formula InChI=1S/C76H52O46/c77-32-1-22(2-33(78)53(32)92)67(103)113-47-16-27(11-42(87)58(47)97)66(102)112-21-52-63(119-72(108)28-12-43(88)59(98)48(17-28)114-68(104)23-3-34(79)54(93)35(80)4-23)64(120-73(109)29-13-44(89)60(99)49(18-29)115-69(105)24-5-36(81)55(94)37(82)6-24)65(121-74(110)30-14-45(90)61(100)50(19-30)116-70(106)25-7-38(83)56(95)39(84)8-25)76(118-52)122-75(111)31-15-46(91)62(101)51(20-31)117-71(107)26-9-40(85)57(96)41(86)10-26/h1-20,52,63-65,76-101H,21H2/t52-,63-,64+,65-,76+/m1/s1
SMILES Oc1c(O)c(O)cc(C(Oc2c(O)c(O)cc(C(OC[C@H]3O[C@H]([C@@H]([C@H]([C@@H]3OC(c4cc(OC(c5cc(O)c(O)c(O)c5)=O)c(O)c(O)c4)=O)OC(c6cc(OC(c7cc(O)c(O)c(O)c7)=O)c(O)c(O)c6)=O)OC(c8cc(OC(c9cc(O)c(O)c(O)c9)=O)c(O)c(O)c8)=O)OC(c%10cc(OC(c%11cc(O)c(O)c(O)c%11)=O)c(O)c(O)c%10)=O)=O)c2)=O)c1
IUPAC Name (2S,3R,4S,5R,6R)-6-(((3,4-dihydroxy-5-((3,4,5-trihydroxybenzoyl)oxy)benzoyl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrakis(3,4-dihydroxy-5-((3,4,5-trihydroxybenzoyl)oxy)benzoate)
Molecular Weight (g/mol) 1701.21
Formula Weight 1700.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.