missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis)

Product Code. 11395798
Change view
Click to view available options
Quantity:
100 x 150 mm
150 x 500 mm
150 x 2000 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395798 150 x 500 mm Each
11395769 100 x 150 mm Each
15495935 150 x 2000 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395798 Supplier Thermo Scientific Alfa Aesar Supplier No. 010351.IV
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Form: approx. 6.3g/100x150mm; Temper: as rolled | CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Quantity 150 x 500 mm
Odor Odorless
Assay Percent Range 99.95% (Metals basis)
Form Foil
Chemical Name or Material Tantalum, Hard

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.