missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane

Product Code. 11349227
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11349227

Brand: Thermo Scientific Alfa Aesar 040038.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16761-83-4 | C15H40O5Ta | 481.428 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16761-83-4
Molecular Formula C15H40O5Ta
Molecular Weight (g/mol) 481.428
MDL Number MFCD00145347
InChI Key KWUQLGUXYUKOKE-UHFFFAOYSA-N
Synonym propan-2-olate,tantalum 5+
PubChem CID 131874823
IUPAC Name propan-2-ol;tantalum
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta]

Specifications

Odor Petrol-like
Linear Formula Ta[OCH(CH3)2]5
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 476.39
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol/hexane
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum(V) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.