missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)

Product Code. 11324587
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11324587

Brand: Thermo Scientific Alfa Aesar 039630.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20219-33-4 | C13H27O6Ta | 460.30 g/mol

Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20219-33-4
Molecular Formula C13H27O6Ta
Molecular Weight (g/mol) 460.30
MDL Number MFCD00145349
InChI Key LSALXRIVOAYYJW-GLUPTGNJSA-M
Synonym tantalum v tetraethoxide 2,4-pentanedionate
PubChem CID 122130355
SMILES [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O

Specifications

Melting Point 45°C
Density 1.5 g/mL
Boiling Point 95°C (0.5 mmHg)
Flash Point >65°C (149°F)
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name ethanol;(E)-4-hydroxypent-3-en-2-one;tantalum
Formula Weight 460.3
Percent Purity 99.99%
Physical Form Solid
Assay (metals basis)
Chemical Name or Material Tantalum(V) tetraethoxide 2,4-pentanedionate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.