missing translation for 'onlineSavingsMsg'
Learn More

Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)

Product Code. 15456765
Change view
Click to view available options
Quantity:
1 m
5 m
10 m
Unit Size:
10m
1m
5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456765 10 m 10m
15436765 1 m 1m
15446765 5 m 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15456765 Supplier Thermo Scientific Alfa Aesar Supplier No. 045133.G6
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Melting Point 2996°C
Boiling Point 5429°C
Density 16.69
Quantity 10 m
Packaging Tube
Formula Weight 180.95
Percent Purity 99.9%
Chemical Name or Material Tantalum wire
What does "hard" mean for the following tantalum wire product: Tantalum wire, 0.203 mm (0.008 in.) dia, hard, 99.9% (metals basis)?

"Hard" tantalum wire refers to the tensile strength, which is >1,000 Mpa.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.