missing translation for 'onlineSavingsMsg'
Learn More

3M™ Tape

Product Code. 12321403
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12321403 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12321403 Supplier 3M™ Supplier No. 7000033143
  • ONLINE EXCLUSIVE PRICE
    412.00 €
    343.29 € / Pack of 1
    Save 68.71 €
    16% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Adhesive Type Acrylate
Color Gray
Product Type Tape
For Use With (Application) For indoor and outdoor use
Length (Metric) 16.5 m
Width (Metric) 19 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.