missing translation for 'onlineSavingsMsg'
Learn More

Taps, TRC

Product Code. 30453640
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30453640 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30453640

Brand: Toronto Research Chemicals TRCT7403051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Taps
CAS 29915-38-6
Molecular Formula C7 H17 N O6 S
Quantity 1 g
Synonym N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid','[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid
InChI Formula InChI=1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14)
SMILES OCC(CO)(CO)NCCCS(=O)(=O)O
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
Molecular Weight (g/mol) 243.278
Formula Weight 243.078
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.