missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq DNA Polymerase

Product Code. 11497020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
3 x 250 U
Unit Size:
750 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497020 3 x 250 U 750 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497020 Supplier MP Biomedicals™ Supplier No. 11EPTQD325
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A specialized thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus

  • Highly processive, recombinant, thermostable 5'-3' DNA polymerase with 10X Buffer with MgCl2 only.
  • Highly reproducible results
  • Ideal for T/A cloning
  • Ultra pure enzyme
  • Highly processive
  • Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands
  • It displays both 5′ to 3′ polymerase and exonuclease activities
  • Quality control performed on every lot of enzymes: Specific PCR on genomic and phage templates, activity check, absence of ribonucleases, absence of nickases and endonucleases, absence of 3' exonucleases, purity on SDS-PAGE, absence of proteases

Specifications

Buffer 10X Buffer with MgCl2
Concentration 5 U/μL
Content And Storage Store at -20°C
For Use With (Application) Nucleic Acid Amplification, PCR, RT-PCR, DNA sequencing and DNA labeling
Includes 20 mM Tris-HCL pH 8.0, 100 mM KCl, 0.1 mM EDTA, 1 mM dithiothreitol, 50% glycerol, 10X buffer with MgCl2
Product Type Proteins, Enzymes & Peptides
Quantity 3 x 250 U
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.