missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Taq DNA Polymerase

Product Code. 16072545
Change view
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16072545 2500 units 2500 units
16062545 1000 units 1000 units
16082545 500 units 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16072545 Supplier Abnova™ Supplier No. D0010.2500Units
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Taq DNA polymerase catalyzes 5'-3' synthesis of DNA.

  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.

Specifications

Concentration 5 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 2500 units
For Use With (Application) Standard PCR
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.