missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Taq-&LOAD™ PCR Mastermix
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 100U
100U
Unit Size:
100 units
1000 units
Description
Taq-&LOAD PCR Mastermix is a 5X concentrated solution containing MP Biomedicals recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer (includes a compound increasing the density of the sample and a red/purple dye). The included red/purple dye migrates at 400 bp on a 1.2% agarose gel.
Specifications
Specifications
| Concentration | 5X |
| Content And Storage | Recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer. Store at -20°C |
| Format | Tube |
| Gel Compatibility | Agarose |
| Label or Dye | Red-purple |
| Quantity | 10 x 100U |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction