missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq Pol Bulk (15U/μL)

Product Code. 11467020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2000U
600U
Unit Size:
2000 units
600 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467020 600U 600 units
11477020 2000U 2000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467020 Supplier MP Biomedicals™ Supplier No. 11EPTQC060
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

Supplied with standard reaction buffer

Provided with QBiogene 10X Incubation PCR Buffer which is suitable for broad range of reaction conditions.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 15 U/μL
Quantity 600U
For Use With (Application) Standard PCR
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.