missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TaqMan™ 2019nCoV Control Kit v1

Product Code. 32096696
Change view
Click to view available options
Ships on dry ice
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32096696 Ships on dry ice
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32096696 Supplier Thermo Scientific™ Supplier No. A47533
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems TaqMan 2019-nCoV Control Kit v1 is a synthetic positive control that contains target sequences for each of the assays included in the TaqMan 2019-nCoV Assay Kit v1.

The Applied Biosystems TaqMan 2019-nCoV Control Kit v1 is a synthetic positive control that contains target sequences for each of the assays included in the TaqMan 2019-nCoV Assay Kit v1 (Cat. No. A47532). The control includes synthetic DNA target sequences for three SARS-CoV-2 genes (ORF1ab, S protein, and N protein) and the human RNase P RPPH1 gene.

During RT-PCR, use 1 μL of the 2019-nCoV Control v1 as the sample for each positive-control reaction.

The control can be used in SARS-CoV-2 research to verify assay performance and to help with troubleshooting.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 tube containing 50 μL (1 x 104 copies/μL)
Store at -10°C to -30°C (Ships on dry ice)
Detection Method Primer-probe
Format Tube
PCR Method 1-step RT-qPCR
Disease COVID-19
Product Line TaqMan
Product Type Covid-19 Detection Kit
Quantity 50 μL (1 x 104 copies/μL)
Shipping Condition Dry Ice
Species Human
Target Organism Class Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
For Use With (Application) Real Time PCR (qPCR)
Volume 1 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.