missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Advanced miRNA Human A and B Cards

Product Code. 16685094 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
754 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16685094 754 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685094 Supplier Applied Biosystems™ Supplier No. A31805
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format.

  • The Applied Biosystems TaqMan Advanced miRNA Human A and B Cards provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format. This product contains two cards, enabling accurate quantitation of 754 unique miRNAs in as little as five hours.
  • Four TaqMan Advanced miRNA Assay endogenous controls are included in each card to aid in data normalization. Details about the included assays are available in the Target List file. The assays can detect and quantify the mature form of the miRNA from 1–10 ng of total RNA from tissue.
  • Required for preparation of the cDNA template that is used with the TaqMan Advanced miRNA assays.
  • The kit enables the analysis of: Multiple miRNAs from a single amplified sample, Samples that are limited in quantity, including serum, plasma, or other biological fluids
  • Universal reverse transcription (RT)—one RT step for all TaqMan Advanced miRNA assays
  • Sensitive—detect as few as 60 copies of input miRNA
  • Specific—detect only mature miRNA and distinguish between highly homologous miRNAs
  • Small sample input—detect and quantify mature miRNA from as little as 1–10 ng of total RNA or 2 μL of plasma or serum
  • Versatile—compatible with tissue and biofluids, including serum and plasma
  • TaqMan Advanced miRNA cards provide all the advantages of TaqMan Advanced miRNA assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan Advanced miRNA assays in parallel. With the TaqMan Advanced miRNA cDNA Synthesis Kit, TaqMan Advanced miRNA cards enable a comprehensive expression profile consistent with miRBase v21, providing the ideal miRNA profiling solution.
  • TaqMan Advanced miRNA cards are compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex.
  • TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal RT chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
  • These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Two 384-well arrays

The TaqMan miRNA cards are shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.
Detection Method Primer-probe
Format 384-well microfluidic card
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast or Standard
Label or Dye 2 Color
Product Line TaqMan
Product Type Advanced miRNA Human A and B Card
Quantity 754 Reactions
Research Category miRNA
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Species Human
For Use With (Application) miRNA analysis
Volume 2 μL
Show More Show Less
What reagents and/or kits are required for the TaqMan® Advanced miRNA Assay workflow using the TaqMan® Advanced miRNA Human A and B cards (Cat. No. A31805)?

All materials required for TaqMan® Advanced miRNA Assay workflow using TaqMan® Advanced miRNA Human and B cards (Cat. A31805) can be found on pages 9-11, in the TaqMan® Advanced miRNA Assays User Guide.

Reagents and kits required for the TaqMan® Advanced miRNA Assay workflow include, but are not limited to:

• RNA Isolation Kit (see recommended kits on page 9)
• TaqMan® Advanced miRNA cDNA Synthesis Kit (Cat. No. A28007)
• TaqMan® Fast Advanced Master Mix (Cat. No. 4444557)

Please refer to the user guide linked above for a full list of other materials and equipment required for the workflow.

I am not seeing good amplification with my TaqMan Advanced miRNA array cards. What is wrong?

The TaqMan Advanced miRNA array cards allow one to profile over 300 different miRNA targets on one card. Due to the large number of targets and the nature of your sample, it is possible that some miRNA on the card will not be expressed on your sample, or expressed at very low levels. This is one reason why you will not see good amplification plots for every spot on the array card. Another possibility is the sample quality or input. Make sure to check the concentration and quality of the RNA sample before reverse transcription. Check the Ct values for your endogenous control assays on the card, and see if it is in the expected range for your typical samples.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.