Learn More
Applied Biosystems™ TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
Shop All Applied Biosystems ProductsDescription
This product contains 1 plate than can be used to evaluate 30 potential endogenous control miRNAs for up to 3 samples. Two exogenous (non-human) controls are included for normalization of sample input.
Features of the TaqMan Advanced miRNA Human Endogenous Controls 96-Well Plate include:
• Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
• Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
• Updated content—assay content updated for mirBase v.21
• Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
• Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families
Product details:
Number of plates: 1
Number of unique miRNAs (not including exogenous controls): 30
Exogenous controls: cel-miR-39-3p, ath-miR159a
Compatible with the following Applied Biosystems qPCR instruments: Viia7, StepOnePlus, 7500/7500 Fast, QuantStudio 3, QuantStudio 5, QuantStudio 6, QuantStudio 7 Flex, and QuantStudio 12k Flex
Recommended master mix: TaqMan Fast Advanced Master Mix
Notes
• TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
• These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Detection Method | Primer-probe |
| Format | 96-Well Plate |
| GC-Rich PCR Performance | High |
| PCR Method | 2-step RT-qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 System, 7900 HT System, QuantStudio™ 12 k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, StepOnePlus™ System |
| Product Line | TaqMan |
| Product Type | Endogenous Control 96-Well Plate |
| Quantity | 1 plate |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.