missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Advanced miRNA Human Endogenous Controls Card

Product Code. 16669382
Change view
Click to view available options
Quantity:
384 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16669382 384 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16669382 Supplier Applied Biosystems™ Supplier No. A34716
  • ONLINE EXCLUSIVE PRICE
    414.00 €
    339.48 € / Pack of 1
    Save 74.52 €
    18% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems TaqMan Advanced miRNA Human Endogenous Controls Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format.

This product contains one card than can be used to evaluate 30 potential endogenous control miRNAs in triplicate for up to four samples. Two exogenous (non-human) controls are included for normalization of sample input.

Features of the TaqMan Advanced miRNA Human Endogenous Controls Card include:
• Reproducibility—pre-spotted assays help reduce pipetting error and minimize experimental variability
• Sample savings—requires only 2 μL purified plasma/serum or 1–10 ng total RNA per sample
• Updated content—assay content updated for mirBase v.22
• Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
• Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

Product details:

Number of cards: 1
Number of unique miRNAs (not including exogenous controls): 30
Exogenous controls: cel-miR-39-3p, ath-miR159a
Compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex
Recommended master mix: TaqMan Fast Advanced Master Mix

Notes
• TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
• These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses a miRNA-specific reverse transcription chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2°C to 8°C and keep protected from light.
Detection Method Primer-Probe Detection
Format 384-well Microfluidic Card
Control Type Endogenous Control
RNAi Type miRNA
Species Human
For Use With (Application) miRNA Analysis
Label or Dye 2 Color
No. of Reactions 384 reactions
Product Line TaqMan
Product Type Endogenous Controls Card
Reaction Speed Fast or Standard
Quantity 384 reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.