Learn More
TaqMan™ Advanced miRNA Human Serum/Plasma 96-well Plates, standard
Shop All Applied Biosystems ProductsDescription
The Applied Biosystems TaqMan Advanced Serum/Plasma 96-Well Plates, Standard, provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 96-well standard plate format. This product contains a set of 2 plates for profiling of up to 188 unique miRNAs in serum/plasma samples, and includes endogenous and exogenous miRNA controls for normalization of data results.
Features of the TaqMan Advanced Serum/Plasma 96-Well Plates include:
- Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
- Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
- Updated content—assay content updated for mirBase v.21
- Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
- Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families
Product details:
- Number of plates: 2
- Number of unique miRNAs (not including exogenous controls): 188
- Exogenous controls: cel-miR-39-3p, ath-miR159a
- Endogenous control: hsa-miR-16-5p
- Compatible with the following Applied Biosystems qPCR instruments: Viia7, 7500, QuantStudio 3, QuantStudio 5, QuantStudio 6, QuantStudio 7 Flex, and QuantStudio 12k Flex
- Recommended master mix: TaqMan Fast Advanced Master Mix
Notes
- TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
- These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Detection Method | Primer-probe |
| Format | 96-Well Plate |
| GC-Rich PCR Performance | High |
| PCR Method | 2-step RT-qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System |
| Product Line | TaqMan |
| Product Type | Advanced miRNA Human Serum/Plasma 96-Well Plate |
| Quantity | 2 Plates |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.