missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ Advanced miRNA Human Serum/Plasma Card

Product Code. 17113099 Shop All Applied Biosystems Products
Click to view available options
Quantity:
384 reactions
Unit Size:
384 reactions
This item is not returnable. View return policy

Product Code. 17113099

Brand: ABI Applied Biosystems A34717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Applied Biosystems TaqMan Advanced Serum/Plasma Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format.

This product contains one card for profiling of up to 188 unique miRNAs in one serum/plasma sample, and includes endogenous and exogenous miRNA controls for normalization of data results.

Features of the TaqMan Advanced Serum/Plasma Card include:

  • Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
  • Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
  • Updated content—assay content updated for mirBase v22
  • Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
  • Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.

These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.
Detection Method Primer-probe
Format 384-well microfluidic card
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast or Standard
Label or Dye 2 Color
Product Line TaqMan
Product Type Advanced miRNA Human Serum/Plasma Card
Quantity 384 reactions
Research Category miRNA
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Species Human
For Use With (Application) miRNA analysis
Volume 2 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.