missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Fast, 96-well Plate; Format 32

Product Code. 15382182 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15382182 1 plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15382182 Supplier Applied Biosystems™ Supplier No. 4413259
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array, Fast, 96-well Plate; Format 32

Custom TaqMan™ Plates are cost-effective and convenient 96-well formats for TaqMan™ Gene Expression Assays. Choose from over 50,000 inventoried human, mouse, rat, canine, and rhesus macaque assays, and from five plate formats - single assays, or assays in duplicate or triplicate.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Fast Plate
Format 96-well Plate, Format 32
Content And Storage Room Temperature
No. of Samples 3
Product Line TaqMan
Quantity 1 plate
Reaction Speed Fast

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.