missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Endogenous Control Panel

Product Code. 11630099 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11630099 2 cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11630099 Supplier Applied Biosystems™ Supplier No. 4367563
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human Endogenous Control Panel

The Applied Biosystems™ TaqMan™ Endogenous Control Array is a 384-well micro fluidic card containing 16 human TaqMan™ Gene Expression Assays commonly used as endogenous controls for for commonly used housekeeping genes and genes that exhibit minimal differential expression across different tissues.

TaqMan™ Endogenous Control Array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Human Endogenous Control Array
Format 384-well Microfluidic Card
Species Human
Content And Storage TaqMan™ Human Endogenous Control Array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file. The cards should be stored at 4°C.
Form Dried Down
Includes Control Primer-Probe Set(s)
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 2 cards
Reaction Speed Standard
Target Gene Housekeeping & Control Gene Panel
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.