missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array Human Endogenous Controls Plate, Fast 96-well
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 96 well plate
Unit Size:
1 set
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Endogenous Control Plate contains 32 genes, plated in triplicate, that have been shown to be good candidates for such an experiment. This collection of genes has been selected from literature searches and⁄or whole genome microarray tests carried out on numerous human tissues. They have been shown to be expressed constitutively and at moderate abundance across most test samples.
Specifications
Specifications
| Content And Storage | Control Primer-Probe Set(s) |
| Form | Dried Down |
| Format | 96-well plate |
| Reaction Speed | Fast |
| Green Features | Sustainable packaging |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Quantity | 1 x 96 well plate |
| Species | Human |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction