missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human GPCR Panel

Product Code. 11650099 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11650099 4 cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11650099 Supplier Applied Biosystems™ Supplier No. 4367785
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human GPCR Panel

The Applied Biosystems™ TaqMan™ Human GPCR Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of human GPCR genes important in drug discovery. This product is designed with 380 TaqMan™ Gene Expression Assays arrayed in singletons in 384 wells (plus 1 control arrayed in quadruplicate) and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include olfactory receptors, retinal receptors, small molecule receptors, chemokine receptors and others including classic endogenous markers. This product is sold in packages of 4 cards.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Array Human GPCR Panel
Format 384-well Microfluidic Card
Species Human
Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Form Dried Down
Includes TaqMan Assay for One 2 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Reaction Speed Standard
Research Category Cell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Development
Target Gene 50 GPCR subfamilies (excludes odorant, olfactory, gustatory and pheromone receptors)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.