missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Human GPCR Pathway, Fast 96-well

Product Code. 10380885 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96-well plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10380885 1 x 96-well plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10380885 Supplier Applied Biosystems™ Supplier No. 4418782
  • ONLINE EXCLUSIVE PRICE
    259.00 €
    212.38 € / 1 sets
    Save 46.62 €
    18% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TaqMan™ Array, Human GPCR Pathway, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human GPCR Pathway 96-well Plate contains 92 assays to genes associated with GPCR pathways and 4 assays to candidate endogenous control genes.
TRUSTED_SUSTAINABILITY

Specifications

Type Array Plate
Format 96-array Plate
Species Human
Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Includes TaqMan Assay for One 10 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 1 x 96-well plate
Reaction Speed Fast
Research Category Immunology, Apoptosis, Biomarker-Related Pathway(s), Signal Transduction, Neurobiology
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.