missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Human Ligand Gated Ion Channels, Fast 96-well
Shop All Applied Biosystems ProductsDescription
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human Ligand Gated Ion Channels Plate targets genes in the 3 superfamilies of the ligand-gated channels: 1) The Cys-loop receptors (GABAA receptor family, Glycine Receptor family, 5-HT Receptors⁄Serotonin, and Nicotinic acetylcholine receptor family); 2) The Ionotropic glutamate receptors (AMPA, Kainate, NMDA); and 3) The ATP-gated Channels (P2X receptors). Also in the panel are genes which encode Ion channel family members: Chloride channel, TRP (transient receptor potential) cation channels, Calcium Channel, and Sodium Channel. Additionally one gene from the Cyclic nucleotide-gated channel and Potassium gated ion channel are also present.
Specifications
Specifications
| Format | 96-array Plate |
| Species | Human |
| Content And Storage | Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. |
| Form | Dried Down |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Quantity | 1 x 96-well plate |
| Reaction Speed | Fast |
| Research Category | Cell Cycle & Proliferation, Signal Transduction, Inflammation |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?