missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human MicroRNA B Cards v3.0

Product Code. 12097789 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Packs
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12097789 4 Packs Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12097789 Supplier Applied Biosystems™ Supplier No. 4444910
  • ONLINE EXCLUSIVE PRICE
    2125.00 €
    1742.50 € / Pack of 4
    Save 382.50 €
    18% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human MicroRNA B Cards v3.0

The TaqMan™ Array Human MicroRNA B Card v3.0 contains 384 TaqMan™ MicroRNA Assays enabling accurate quantitation of 377 human microRNAs in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human is included as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool B v3.0 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool B v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Human MicroRNA B Cards v3.0
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Real-time PCR (qPCR), miRNA and non-coding RNA analysis
Quantity 4 Packs

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.