missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Human Molecular Mechanisms of Cancer, Fast 96-well

Product Code. 10699075 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10699075 1 Ea. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10699075 Supplier Applied Biosystems™ Supplier No. 4418806
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TaqMan™ Array, Human Molecular Mechanisms of Cancer, Fast 96-well

These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan™ Array Human Molecular Mechanisms Of Cancer 96-well Plate contains 92 assays to genes accociated with the molecular mechanisms of cancer and 4 assays to candidate endogenous control genes.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Human Molecular Mechanisms of Cancer
Format 96-array Plate
Content And Storage 4°C
For Use With (Application) Oncology and genetic disease research, pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
Includes TaqMan™ array plate well contains a single TaqMan™ assay for one 10μL reaction
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.