missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Nuclear Receptors Panel

Product Code. 11610119 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11610119 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11610119 Supplier Applied Biosystems™ Supplier No. 4379961
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human Nuclear Receptors Panel

The Applied Biosystems™ TaqMan™ Human Nuclear Receptor Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 64 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.

The nuclear hormone receptor family is highly conserved in vertebrates. They have a modular domain structure consisting of a DNA-binding domain and a ligand-binding domain. Other domains include a modulatory A⁄B domain, the hinge D domain, and a variable C-terminal F domain that is missing in some nuclear hormone receptors. The TaqMan™ Nuclear Receptor Gene Signature Array contains 48 human nuclear hormone receptors (and 16 controls) and has been classified according to sequence homology into seven subfamilies. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Human Nuclear Receptors Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.