missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Mouse Endogenous Control Panel

Product Code. 11600109 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11600109 2 cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11600109 Supplier Applied Biosystems™ Supplier No. 4378702
  • ONLINE EXCLUSIVE PRICE
    1050.00 €
    861.00 € / Pack of 1
    Save 189.00 €
    18% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Mouse Endogenous Control Panel

The Applied Biosystems™ TaqMan™ Mouse Endogenous Control Array is a 384-well micro flluidic card containing 16 mouse TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Array Mouse Endogenous Control Panel
Format 384-well Microfluidic Card
Species Mouse
Content And Storage TaqMan™ Mouse Endogenous Control Array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file. The cards should be stored at 4°C.
Form Dried Down
Includes Control Primer-Probe Set(s)
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 2 cards
Reaction Speed Standard
Target Gene Housekeeping & Control Gene Panel
Visa mer Visa mindre

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.