missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Mouse Stem Cell Pluripotency Panel

Product Code. 11630119 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11630119 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11630119 Supplier Applied Biosystems™ Supplier No. 4385363
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Mouse Stem Cell Pluripotency Panel

The Applied Biosystems™ TaqMan™ Mouse Stem Cell Pluripotency Array is an efficient, easy-to-use micro fluidic card for the characterization of undifferentiated stem cells or their differentiated derivatives. This gene signature panel contains mouse orthologs of well-defined human markers for pluripotency plus 6 endogenous controls. The format of the TaqMan Array allows for 1-4 replicates to be run in parallel per array. Each order includes a CD with assay information and an instrument software input file. This product is sold in packages of 4 cards.

The Mouse Stem Cell Pluripotency Array contains mouse ortholog genes of the human panel to characterize mouse embryonic stem cells. The gene content on this TaqMan Array resulted directly from the work of the I.S.C.I consortium and characterization of human embryonic stem cell lines by the International Stem Cell Initiative.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

The Right Targets

These TaqMan Arrays use Applied Biosystem’s proprietary micro fluidics technology to simultaneously analyze gene expression levels by quantitative PCR for 92 test genes and 6 endogenous controls. Genes were included based on the following criteria:
    Expression in human embryonal carcinoma cells (hEC), primordial germ cells or human ES (hES) cells
    Known role in maintenance of pluripotency in hES cells
    Identification in microarray-based experiments as being correlated with the stem cell state
    Expression known to be characteristic of specific differentiation lineages in the post implantation embryo and mature tissues. These are the right targets to characterize embryonic stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Mouse Stem Cell Pluripotency Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Real-time PCR (qPCR), real-time PCR-based gene expression profiling, stem cell analysis
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.