missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Rat GPCR Panel

Product Code. 11670109 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11670109 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11670109 Supplier Applied Biosystems™ Supplier No. 4378709
  • ONLINE EXCLUSIVE PRICE
    2450.00 €
    2009.00 € / Pack of 1
    Save 441.00 €
    18% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Rat GPCR Panel

The Applied Biosystems™ TaqMan™ Mouse GPCR Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of mouse GPCR The TaqMan™ Rat GPCR Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of rat GPCR genes important in drug discovery. This product is designed with 380 TaqMan™ Gene Expression Assays arrayed in singletons in 384 wells (plus 1 control arrayed in quadruplicate) and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include retinal receptors, small molecule receptors, chemokine receptors and others including classic endogenous control targets. GPCR Panels are sold in packs of 4.

The GPCR targets selected fall into the 50 subfamilies listed, many of which are drug targets or disease associated —odorant, olfactory, gustatory or pheromone receptors are not included. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Rat GPCR Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Cell viability, proliferation and function, pharma and biopharma, real-time PCR (qPCR), target and lead identification and validation
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.