missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Rat Inflammation Panel

Product Code. 11660109 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11660109 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11660109 Supplier Applied Biosystems™ Supplier No. 4378708
  • ONLINE EXCLUSIVE PRICE
    2140.00 €
    1754.80 € / Pack of 1
    Save 385.20 €
    18% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Rat Inflammation Panel

The Applied Biosystems™ TaqMan™ Rat Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and the panel is sold in packs of 4.

The TaqMan™ Gene Signature Arrays for rat inflammation contain over 90 genes that are, or have been, studied as targets for a range of inflammatory diseases. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Array Rat Inflammation Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.