missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Rat Inflammation

Product Code. 10168014 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 standard plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10168014 1 standard plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10168014 Supplier Applied Biosystems™ Supplier No. 4414081
  • ONLINE EXCLUSIVE PRICE
    267.00 €
    218.94 € / 1 sets
    Save 48.06 €
    18% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Rat Inflammation

The panel of assays in this plate targets genes involved in a broad range of inflammatory diseases. The genes fall into 4 classes:

  • Channels (L-type calcium and Ligand gated);
  • Enzymes and Inhibitors (Lipases, Kinases, Nitric Oxide Synthase, Phosphodiesterase, Prostaglandin Metabolism, and Proteases)
  • Factors (Tumor necrosis factors, Nuclear factor Kappa-Β, Interleukin, Annexins, Kinnogens)
  • Receptors (GPCR, IL Receptor family, Adhesion molecules, TNF receptors and Nuclear receptors)

    Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate Applied Biosystems™ TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.

  • Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
  • Fast delivery; on your bench in days
  • Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
  • Same TaqMan™Assay Quality, New Format

    TaqMan™ Array Plates contain the same high quality TaqMan™ Gene Expression Assays (PCR primers and TaqMan™ probe sets) that have been commercially available for years in a convenient, dried, 96-well format. Extensive tests have found no difference in performance between TaqMan™Assays supplied in the traditional liquid format and assays dried into a TaqMan™ Array Plate. Figure 1 shows the results of an experiment comparing the linear range of the two formats, where no difference in performance was observed.

    Just Add Master Mix and Sample, and Begin Cycling
    Ideal for projects such as validation of microarray and RNAi data, and pathway studies, the plates include just the right amount of TaqMan™Assay in each well: enough for one 20μl reaction. You can order as few as three Gene Signature Plates, making these an affordable option for many laboratories and minimizing the waste of unused individually purchased TaqMan™Assays. Save a significant amount of time setting up your experiments. Just add master mix and sample, and begin cycling.

    Figure 2 is an example Plate Layout with Gene Symbols and Assay IDs

    Plate Map Ensures Proper Data Labeling and Easy Re-Ordering
    Each pack of TaqMan™ Array Plates is shipped with a CD containing the plate configuration map (see Figure 2) along with the details of each TaqMan™Assay. This information is imported into the real-time PCR instrument to enable proper labeling of experimental data and easy access to Assay ID numbers for simple reordering.

    Simple to Use
    Each TaqMan™ Array Plate well contains a single TaqMan™Assay for one 20μl reaction. The plates can be stored at room temperature or at 4°C. When you are ready to run your assays, simply add PCR master mix and your cDNA sample, seal the plate, and begin cycling on a real-time PCR instrument.

    Compatibility
    TaqMan™ Array Plates have been optimized for use with TaqMan™ Gene Expression Master Mix, but are also compatible with TaqMan™ Universal PCR Master Mix. For studies using cultured cells, TaqMan™ Array Plates are also compatible (and recommended) with the TaqMan™ Gene Expression Cells-to-Ct™ Kit. TaqMan™ Array Plates must be run on a real-time PCR instrument with 96-well plate capability. The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μl reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type Array Plate
    Format 96-array Plate
    Species Rat
    Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
    Form Dried Down
    Includes TaqMan Assay for One 20 uL Reaction
    Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
    Label or Dye FAM
    Product Line TaqMan
    Purification Method Standard
    Quantity 1 standard plate
    Reaction Speed Standard
    Research Category Inflammation, Signal Transduction
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.