missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Rodent MicroRNA B Cards v3.0

Product Code. 12077789 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Packs
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12077789 4 Packs Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12077789 Supplier Applied Biosystems™ Supplier No. 4444899
  • ONLINE EXCLUSIVE PRICE
    2110.00 €
    1730.20 € / Pack of 4
    Save 379.80 €
    18% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Rodent MicroRNA B Cards v3.0

The TaqMan™ Array Rodent MicroRNA B Card v3.0 contains 384 TaqMan™ MicroRNA Assays enabling accurate quantitation of 306 and 135 unique microRNAs for mouse and rat respectively in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls for each species are included to aid in data normalization and one TaqMan MicroRNA Assay not related to rodent is included as a negative control. Detail on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Rodent Pool B v3.0 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Rodent Pool B v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.

TaqMan™ Array MicroRNA Cards provide all the advantages of TaqMan™ MicroRNA Assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan™ MicroRNA Assays in parallel.

Megaplex™ RT Primers are highly multiplexed RT primers designed to convert up to 381 miRNAs to cDNA prior to real-time analysis, and are required to run the TaqMan™ Array MicroRNA Card. For greater sensitivity, or when working with limited sample, Megaplex™ PreAmp Primers provide an optional preamplification step prior to real-time analysis. TaqMan™ Array MicroRNA Cards and Megaplex™ Primer Pools enable a comprehensive expression profile consistent with miRBase v21 to be created in as little as 5 hours, providing the ideal miRNA profiling solution. Key product features:

Highly specific—quantitate only the biologically active mature miRNAs

  • Highly streamlined workflow reduces the number of RT (reverse transcription) reactions per sample
  • Simplify sample handling and increase sample throughput
  • Coverage for human, mouse and rat species
  • An ideal miRNA profiling solution

    Simplify Sample Handling and Increase Sample Throughput
    Following reverse transcription of miRNA targets using Megaplex™ RT Primers and optional preamplification using Megaplex™ PreAmp Primers, TaqMan™ Universal PCR Master Mix II is simply combined with each reaction and pipetted into each sample loading port in the TaqMan™ Array Card, simplifying sample handling and increasing sample throughput.

    Provides Comprehensive Coverage of Sanger miRBase
    Megaplex™ RT Pools and PreAmp Primers are available for human and rodent species, and consist of pre-selected and matched content across a series of primer pools and TaqMan™ Array Cards. The content is derived from miRBase v14 and v15 for human and rodent species respectively, providing close to full coverage of Sanger with the most up-to-date TaqMan™ MicroRNA Assays.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type TaqMan™ Array Rodent MicroRNA B Cards v3.0
    Format 384-well Microfluidic Card
    Content And Storage 2°C to 8°C
    For Use With (Application) Real-time PCR (qPCR), miRNA and non-coding RNA analysis
    Includes 384-well arrays and information CD
    Quantity 4 Packs

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.