missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Cells-to-CT™ HepatoExpress™ Kit

Product Code. 31722752
Change view
Click to view available options
No. of Reactions:
100 Reactions
2500 Reactions (Lysis only)
40 Reactions
400 Reactions
Unit Size:
1 set
100 reactions
40 reactions
400 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
31722752 400 Reactions 400 reactions
31722965 40 Reactions 40 reactions
31722703 100 Reactions 100 reactions
31722757 2500 Reactions (Lysis only) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31722752 Supplier Invitrogen™ Supplier No. A65790
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TaqMan Cells-to-CT HepatoExpress Kit is a complete cell lysate system designed for gene expression analysis directly from hepatocytes without the need of RNA purification.

The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a hepatocyte gene expression kit designed for RNA extraction-free RT-qPCR. It enables fast, direct lysis of primary hepatocytes, eliminating the need for RNA purification while reducing sample preparation time to ~10 minutes.

This streamlined cell-to-CT workflow allows researchers to perform high-sensitivity gene expression analysis directly from cells, without columns, beads, or centrifugation. The result is reliable, reproducible RT-qPCR data with significantly reduced hands-on time.

Optimized specifically for primary hepatocyte models—including ECM-overlay cultures—the kit supports high-throughput, automation-compatible workflows for drug metabolism, toxicity studies, and mechanistic research.

Key Benefits

Fast, Extraction-Free Gene Expression Workflow

  • Perform RT-qPCR without RNA isolation or purification
  • Reduce RNA preparation time from hours to ~10 minutes
  • Direct lysis workflow minimizes sample handling and variability

High-Quality RT-qPCR Performance

  • Sensitivity and reproducibility comparable to purified RNA workflows
  • Preserves RNA integrity during lysis
  • Compatible with multiplex qPCR and low-input samples

Optimized for Primary Hepatocytes

  • Efficient lysis of hepatocytes, including ECM-overlay cultures
  • Consistent performance across hepatocyte species and models
  • Designed for challenging in vitro liver systems

Automation-Ready and Scalable

  • Compatible with 96-well plate formats and robotic liquid handling systems
  • An ideal choice for high-throughput screening and large-scale studies
  • Room-temperature workflow simplifies lab integration

Sustainable and Simplified Workflow

  • Significantly reduces plastic consumables associated with RNA purification
  • REACH-compliant reagents support environmentally responsible lab practices

Designed for Hepatocyte Gene Expression Challenges

Primary hepatocytes are widely used for drug metabolism, CYP450 induction, and toxicity studies, but efficient RNA extraction from these cells—especially in ECM-overlay cultures—can be difficult and time-consuming.

The HepatoExpress Kit addresses these challenges with a direct lysis chemistry that:

  • Effectively penetrates extracellular matrix (ECM) overlays
  • Protects RNA during lysis without purification
  • Maintains compatibility with reverse transcription and qPCR

This enables accurate gene expression analysis without the variability introduced by traditional RNA extraction workflows.

Applications

The HepatoExpress Kit is an ideal choice for:

  • Hepatocyte gene expression analysis
  • CYP450 induction assays
  • Drug metabolism and ADME studies
  • Toxicology and safety screening
  • Target verification and early-stage compound screening
  • Mechanistic studies in liver biology

Scalable from Discovery to High-Throughput Screening

Whether performing small-scale mechanistic studies or large-scale screening, the kit supports flexible experimental design. Its automation-friendly format enables seamless integration into high-throughput workflows, improving reproducibility and throughput.

Workflow: From Cells to qPCR in ~75 Minutes

The simple three-step workflow enables rapid gene expression analysis:

  1. Cell lysis (~10 minutes, room temperature)
    • Simultaneous cell lysis, RNA preservation, and genomic DNA removal
  2. Reverse transcription
    • cDNA synthesis using SuperScript™ IV VILO™ Master Mix
  3. qPCR detection
    • Sensitive, multiplex-capable amplification using TaqMan™ Gene Expression Assays

Compatible with 700,000+ TaqMan™ Gene Expression Assays, enabling broad pathway and target coverage.

Why Choose an RNA Extraction-Free qPCR Workflow?

Compared to traditional column-based RNA isolation methods, the HepatoExpress Kit:

  • Reduces workflow time from hours to minutes
  • Minimizes sample loss and variability
  • Reduces multiple handling steps that can degrade RNA
  • Improves consistency across replicates and experiments

The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a fast, reliable, and scalable solution for primary hepatocyte gene expression analysis. By enabling direct, RNA extraction-free RT-qPCR, it simplifies workflows while maintaining high data quality—making it excellent for modern drug discovery and in vitro liver research.

Addition to HepatoExpress Product Description:

In addition to the applications listed above, the HepatoExpress Cells-to-CT Kit may be suitable for use with 3D culture systems based on customer-reported experience across a range of experimental models. These reports suggest that the kit can be incorporated into workflows involving complex cellular architectures, such as spheroids or organoid cultures, without the need for extensive protocol modification. However, this application has not been independently validated by Thermo Fisher Scientific and is not included within the kit's standard performance specifications. As such, users should evaluate suitability within their specific system and experimental conditions, as results may vary depending on the culture model and workflow.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage HepatoExpress Lysis Solution (Room Temperature), HepatoExpress LysisGuard (-20°C), Express ezDNase (-20°C), SuperScript IV VILO Master Mix (-20°C), SuperScript IV VILO No RT Control (-20°C), TaqMan Fast Advanced Master Mix (4°C). For long-term storage, store HepatoExpress Lysis Buffer at 4°C.
Detection Method TaqMan
Form Liquid
Format Kit
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Reaction Speed 10 min.
High-throughput Compatibility Yes
Label or Dye ROX
Product Type Direct Cell Lysis to RT-qPCR Kit
Quantity 400 Reactions
Shipping Condition Frozen, Dry Ice
Target cDNA
For Use With (Application) Gene Expression
Isolation Technology Direct Cell Lysis
No. of Reactions 400 Reactions
Purification Target RNA
Sample Type Hepatocytes, HepaRG, HepG2
Starting Material Up to 100,000 Cultured Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.