missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ EGFR Exon 19 Deletions Assay

Product Code. 15574354 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
75 reactions
Unit Size:
150µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15574354 75 reactions 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574354 Supplier Applied Biosystems™ Supplier No. 4465805
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ EGFR Exon 19 Deletions Assay

TaqMan™ Mutation Detection Assays detect somatic mutations in genes that are associated with cancer. These assays are powered by competitive allele-specific TaqMan™ PCR (castPCR™) technology. Specific and sensitive, castPCR™ can detect rare mutated DNA among wild-type DNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description TaqMan™ EGFR Exon 19 Deletions Assay, 150 μL, Designed to Address these Limitations, as well as to Provide a Method for Accurately Quantifying the Percentage of Specific Cancer Mutations within a Sample
Detection Method Primer-probe
PCR Method qPCR
Product Line TaqMan
Product Type TaqMan™ EGFR Exon 19 Deletions Assay
Quantity 75 reactions
Volume 150 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.