missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0

Product Code. 10704407 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
200 Reactions
400 Reactions
800 Reactions
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10704407 400 Reactions Each
10575575 200 Reactions Each
10085544 800 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10704407 Supplier Applied Biosystems™ Supplier No. 4398021
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors.

Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0 offers accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors. Use this Master Mix to analyze environmental, food and other challenging samples.

  • Tailored for quantitative, real-time PCR experiments
  • Unrivalled sensitivity for challenging applications, low copy number target detection and multiplex PCR
  • Includes pathogen detection and viral⁄bacterial load quantification
  • Sensitive detection for reliable quantification of abundant and limited genetic targets
  • Stable mix for high-throughput handling
  • Upon receipt, store at -15°C to -25°C before first use, protected from light
  • After first use, store at 2°C to 8°C, protected from light

Optimized Formulation for Unrivalled Performance

TaqMan Environmental Master Mix 2.0 is a convenient 2X mix for target quantification that includes:

  • AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure), a highly purified DNA Polymerase for improved detection of bacterial targets. This hot-start enzyme is inactive at room temperature so reactions can be set up on the benchtop. Enzyme is activated during thermal cycling.
  • Passive internal reference based on proprietary ROX™ dye for precise data analysis.

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage This TaqMan™ Environmental Master Mix 2.0 is a convenient 2X mix in a 10 mL bottle that includes: AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), and a passive internal reference based on proprietary ROX dye for precise data analysis.
Upon receipt, store at -15 to -25°C before first use, protected from light. After first use, store at 2 to 8°C, protected from light.
Detection Method Primer-probe
Format Bottle
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type qPCR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 400 reactions
Sufficient For 400 Reactions
For Use With (Application) Gene Expression, Genotyping, Pathogen Detection, Virus Detection
Hot Start Built-In Hot Start
No. of Reactions 400 Reactions
Sample Type Environmental Sample, Food
Volume 10 mL
Show More Show Less
Is the 2X Environmental Master Mix the same as the TaqMan Universal PCR Master Mix, with or without UNG?

No, the 2X Environmental Master Mix (EMM) is not the same as the TaqMan Universal PCR Master Mix (with or without AmpErase UNG). The 2X Environmental Master Mix is specifically formulated to detect specific bacterial pathogens with greater specificity and sensitivity. It includes AmpliTaq Gold DNA Polymerase, LD, which is a highly purified version of AmpliTaq Gold DNA Polymerase. Every effort has been made in the development of this new polymerase to eliminate bacterial DNA from the enzyme preparation.

Is the TaqMan Environmental Master Mix 2.0 (Cat. No. 4398021, 4398044, 4396838) compatible with the QuantStudio instruments?

Yes, it is.

I have run out of Environmental Master Mix v2.0 from the Ion 16S Metagenomics Kit (Cat. No. A26216). Do you offer this as a standalone item?

Yes. You can purchase the TaqMan Environmental Master Mix 2.0 (Cat. No. 4396838).

Is there a mastermix that I can use with fecal samples?

For fecal samples, we recommend a master mix that is very tolerant of inhibitors, such as our TaqMan Environmental Master Mix 2.0. Please refer to the following paper: https://assets.thermofisher.com/TFS-Assets/CMD/posters/cms_089094.pdf

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.