missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Gene Expression Assays

Product Code. 11967021 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
250 Reactions, Inventoried
2900 Reactions, Made to Order
360 Reactions, Made to Order
75 Reactions, Inventoried
75 Reactions, Made to Order
750 Reactions, Made to Order
L (2900 Reactions), Made to Order
M (750 Reactions), Made to Order
S (360 Reactions), Made to Order
Label or Dye:
ABY
Cy5
FAM
JUN
Unit Size:
360µL
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Label or Dye Unit Size
11967021 75 Reactions, Made to Order FAM Each
10794597 250 Reactions, Inventoried FAM Each
11997011 2900 Reactions, Made to Order FAM Each
11620099 750 Reactions, Made to Order FAM Each
10330545 360 Reactions, Made to Order FAM 360µL
11977021 75 Reactions, Inventoried FAM Each
32044651 S (360 Reactions), Made to Order FAM Each
32044894 M (750 Reactions), Made to Order FAM Each
32045134 L (2900 Reactions), Made to Order FAM Each
32226451 S (360 Reactions), Made to Order ABY Each
32226518 M (750 Reactions), Made to Order ABY Each
32045072 L (2900 Reactions), Made to Order ABY Each
32226608 S (360 Reactions), Made to Order JUN Each
32226471 M (750 Reactions), Made to Order JUN Each
32044719 L (2900 Reactions), Made to Order JUN Each
32226612 S (360 Reactions), Made to Order Cy5 Each
32226620 M (750 Reactions), Made to Order Cy5 Each
32044728 L (2900 Reactions), Made to Order Cy5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 11967021 Supplier Applied Biosystems™ Supplier No. 4448892
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM, VIC, ABY, JUN, and Cyanine 5) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Features of TaqMan Gene Expression assays include:

  • Easy to use—just add cDNA and master mix and run—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Extensive content—Over 1.8 million predesigned assays available for over 25 different species
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Fast delivery—your assays shipped in as little as 1–2 days (see below)
  • Multiplexing ready—TaqMan assays are now available with 5 different dye labels to support simultaneous detection of up to 5 targets in a single qPCR reaction. FAM, ABY, JUN, and Cyanine 5 assays are available with balanced primer/probe concentrations (Primer-Limited, 350 nM) to support consistent multiplex performance. VIC Primer-Limited assays (150 nM) are available with SKU 4448484 (S), 4448485 (M), 4448486 (L).

Approximate ship time

  • Inventoried: 1–2 days in North America, 3–5 days in Europe*
  • Made to order: 4–6 days in North America, 6–10 days in Europe (FAM, VIC); 4-10 days in North America, 6-14 days in Europe (ABY, JUN, Cyanine 5)

TaqMan Gene Expression assays are the gold standard in qPCR and dPCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Pre-formulated assay

  • 2 unlabeled PCR primers (Standard: 900 nM each final 1X concentration; Primer-Limited: 350 nM each final 1X concentration)
  • 1 dye-labeled (FAM, VIC, ABY, JUN or Cyanine 5) TaqMan MGB probe (250 nM final 1X concentration)

All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays Guarantee.**

*Our most popular assays are stocked in North America and Europe for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556) or TaqPath DuraPlex 1-Step RT-qPCR Master Mix (Cat. No. A58666)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Content And Storage 1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15°C to -25°C.

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Label or Dye FAM
Product Line TaqMan
Product Type Gene Expression Assay
Quantity 75 Reactions, Made to Order
Shipping Condition Room Temperature
Fidelity (vs. Taq) 60 X
No. of Reactions 75 Reactions, Made to Order
Show More Show Less
compliance-icons
Product Identifier
  • TAQMAN GENE EX ASSAYS MTO
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.