missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ GMO Screening Kit

Product Code. 13592540 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
192 Reactions
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
13592540 192 Reactions Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13592540 Supplier ABI Applied Biosystems Supplier No. 4466334
  • ONLINE EXCLUSIVE PRICE
    1308.33 €
    1257.99 € / Pack of 4
    Save 50.34 €
    3% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ GMO Screening Kit

The TaqMan™ GMO Screening Kit analyzes DNA extracted from food and feed samples for the presence of the main transgenic regulatory regions. This real-time PCR kit contains all reagents (except for the template DNA) required for the detection of genetically modified organisms (GMOs ). In addition, the kit contains an Internal Positive Control (IPC) to identify possible PCR inhibition. The benefits of the Taqman™ GMO Screening Kit over other commercially available products include:

  • Flexibility—GMOs detected in DNA obtained from ANY food or feed source
  • Accuracy—an internal positive control (IPC) rules out inhibition during qPCR process
  • Speed—results provided in less than 3 hours

    The TaqMan™ GMO Screening Kit is capable of multiplex amplification of Endogenous Vegetal Control, Internal Positive Control, and these regulatory regions:
     
  • P35S/CaMV
  • TNOS/A. tumefaciens
  • P34S/FMV

    All of which make possible the detection of significant events such as soybean MON89788.

    The TaqMan™ GMO Screening Kit provides all of the necessary reagents for testing 48 samples and is optimized for use in all Applied Biosystems™ real-time PCR instruments. And because it is a real-time PCR kit, no electrophoresis is required. In addition, the kit offers extremely high sensitivity, detecting targets with only 5 DNA copies.

    The development and manufacture of this kit occurs through a partnership with IMEGEN, which has a long history of GMO research and development.

    Other related products include:
    Taqman™ Roundup Ready Soya Quantification Kit
    GMO Extraction Kit
    TaqMan™ GMO Maize Quantification Kit
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains all reagents (except for the template DNA) required for the detection of genetically modified organisms (GMOs ). Internal Positive Control (IPC) to identify possible PCR inhibition
Description TaqMan™ GMO Screening Kit, Analyzes DNA Extracted from Food and Feed Samples for the Presence of the Main Transgenic Regulatory Regions
Detection Method Primer-probe
Kit Type TaqMan™ GMO Screening Kit
Product Type TaqMan™ GMO Screening Kit
Target P35S, TNOS, P34S and Vegetal control gene
No. of Reactions 192 Reactions
Sample Type Food

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.