Learn More
Applied Biosystems™ TaqMan™ Mutation Detection Assays
Product Code. 15205624 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
150 μL
Unit Size:
10 units
Product Code. Quantity Unit Size
15205624 150 μL 10 units
1 options
Product Code. 15205624 Supplier Applied Biosystems™ Supplier No. 4465804
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
TaqMan Mutation Detection Assays
TaqMan™ Mutation Detection Assays detect somatic mutations in genes that are associated with cancer. These assays are powered by competitive allele-specific TaqMan™ PCR (castPCR™) technology. Specific and sensitive, castPCR™ can detect rare mutated DNA among wild-type DNA.
TRUSTED_SUSTAINABILITY
Specifications
Description TaqMan™ Mutation Detection Assays, 150μL, Designed to Address these Limitations, as well as to Provide a Method for Accurately Quantifying the Percentage of Specific Cancer Mutations within a Sample
Product Type TaqMan™ Mutation Detection Assays
Quantity 150 μL