Learn More
Applied Biosystems™ TaqMan™ OpenArray™ Genotyping Plate, Custom Format 128 QuantStudio™ 12K Flex
TaqMan™ OpenArray™ Genotyping Plate, Custom Format 128 QuantStudio™ 12K Flex
Brand:  Applied Biosystems™ 4471116
Product Code. 12827432
  • 6860.00 € / Pack of 1
Description
Design your own PCR plate, building from custom assays and over 4.5 million predesigned genotyping assays, including 2,700 inventoried Drug Metabolizing Enzyme (DME) assays. The subsequent qPCR system workflow facilitates the validation/screening of up to 256 SNPs across >1,500 samples per day.