missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR)

Product Code. 15344674 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
200 Reactions
40 Reactions
Unit Size:
200 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15344674 200 Reactions 200 reactions
10515405 40 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15344674 Supplier Applied Biosystems™ Supplier No. 4488593
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ PreAmp Master Mix pre-amplifies small amounts of cDNA without introducing amplification bias to the sample.

TaqMan™ PreAmp Master Mix lets you: • Analyze mRNA from precious samples such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE) • Stretch as little as 1 ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression assays • Analyze up to 100 gene expression targets with minimal hands-on time Preserves Equilibrium of Targets Without Preamplification Bias: In the past, preamplification kits and techniques often resulted in uneven amplification of targets and incorrect or biased data (as shown by low correlation coefficients between preamplified and unamplified samples). TaqMan™ PreAmp Master Mix amplifies with no bias and provides extremely high correlation between amplified and unamplified cDNA. Easy Workflow: With TaqMan™ PreAmp Master Mix, preamplification setup takes only a few minutes. After a one-time pooling of the TaqMan™ assays , preamplification setup (mixing the pooled assays with cDNA sample and TaqMan™ PreAmp Master Mix) takes only five additional minutes. Cycling takes 1.5 hours using a GeneAmp™ 9700 thermal cycler. For Research Use Only. Not for use in diagnostic procedures.

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one 5-mL bottle of 2X master mix.

Store at 2–8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
Format Bottle
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
Product Line TaqMan
Product Type PreAmp Master Mix
Quantity 200 reactions
Sufficient For 200 Reactions
For Use With (Application) Pre-amplification
No. of Reactions 200 Reactions
Sample Type cDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.