missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ RNase P Assay, ABY™ dye/QSY™ probe

Product Code. 15611812 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
250 Reactions
Unit Size:
250 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15611812 250 Reactions 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15611812 Supplier Applied Biosystems™ Supplier No. 4485714
  • ONLINE EXCLUSIVE PRICE
    327.00 €
    268.14 € / 250 reactions
    Save 58.86 €
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Enables relative gene expression quantification in cDNA samples when used with other gene expression assays

  • The Applied Biosystems™ TaqMan™ RNaseP Assay with ABY™ dye-labeled QSY™ probe provides a pre-formulated assay for quantitating human RNase P. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays.
  • It consists of an ABY™ dye-labeled QSY™ probe plus sequence-specific forward and reverse primers, and it can be used for multiplex or singleplex PCR reactions.
  • Eliminate assay design time using pre-designed primers and pre-designed ABY™ dye-labeled TaqMan™ probe
  • Minimize experimental optimization time
  • Use in conjunction with FAM™ or VIC™ dye-labeled predesigned gene expression assays
  • Save time in assay design, formulation, and testing by using the TaqMan™ RNase P Assay as your control when quantifying gene expression
  • This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.
  • The ABY™ dye is optimized for use with the 3rd filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NR_002312.1
Amplicon Size 87
Content And Storage Primer-Probe Set(s)
Detection Method Primer-probe
Format Tube
PCR Method qPCR
Gene Symbol RNase P (RPPH1)
Internal Probe Modification QSY™ Non-Fluorescent Quencher (3')
Label or Dye ABY
Product Line TaqMan
Product Type Real Time PCR Reagents
Quantity 250 Reactions
Species Human
Target Ribonuclease P RNA component H1
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 250 Reactions
Show More Show Less
What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?

ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.

Please refer to your instrument user guide for additional information on compatibility with different dye channels.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.