missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ RNase P Assay, VIC™ dye/QSY™ probe, primer-limited

Product Code. 17168705 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
250 reactions
Unit Size:
250 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17168705 250 reactions 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17168705 Supplier Applied Biosystems™ Supplier No. a30065
  • ONLINE EXCLUSIVE PRICE
    358.00 €
    293.56 € / 250 reactions
    Save 64.44 €
    18% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ RNase P Assay, VIC™ dye/QSY™ probe, primer-limited

  • The Applied Biosystems™ TaqMan™ RNase P Assay with VIC™ dye-labeled QSY™ probe provides a pre-formulated assay for quantitating human RNase P. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. It consists of a VIC dye-labeled QSY probe plus sequence-specific forward and reverse primers. The assay can be used for multiplex or singleplex PCR reactions and it is primer limited, so it can be used with samples that overexpress RNase P. For a primer-limited assay with ABY™ dye-labeled probe rather than VIC dye-labeled probe, see Cat. No. 4485715.
  • Eliminate assay design time using pre-designed primers and pre-designed VIC dye-labeled TaqMan probe
  • Minimize experimental optimization time
  • Use in conjunction with FAM™ dye-labeled predesigned gene expression assays
  • Formulated at a primer-limited concentration for use in multiplex reactions
  • Save time in assay design, formulation, and testing by using the TaqMan RNase P Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.
  • In order to facilitate multiplex experiments, the assay formulation is primer limited, allowing the assay to be used with high RNase P expressors.
  • The VIC dye is optimized for use with the second filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in Freezer (-5°C to -30°C).
For Use With (Equipment) Real Time PCR System
Product Line QSY, TaqMan, VIC
Product Type Rnase P Assay
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Approved for Shipment at Room Temperature or on Wet Ice
Quantity 250 reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.