missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Rodent MicroRNA A Array v2.0

Product Code. 12073719 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Packs
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12073719 4 Packs Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12073719 Supplier Applied Biosystems™ Supplier No. 4398967
  • ONLINE EXCLUSIVE PRICE
    2155.00 €
    1767.10 € / Pack of 4
    Save 387.90 €
    18% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Rodent MicroRNA A Array v2.0

The Applied Biosystems™ TaqMan™ Rodent MicroRNA A Array v2.0 contains 384 TaqMan™ MicroRNA Assays enabling accurate quantitation of 335 and 226 unique microRNAs for mouse and rat respectively in as little as 5 hours. Three TaqMan™ MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human is included as a negative control. Details of the included assays is provided in the Array Map file. Purchase of the Megaplex™ RT Primers, Rodent Pool A is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Rodent Pool A for situations where sensitivity is of the utmost importance and/or sample is limiting.

TaqMan™ MicroRNA Arrays
provide all the advantages of TaqMan™ MicroRNA Assays in a convenient, pre-configured micro fluidic card minimizing experimental variability and effort required to run 384 TaqMan™ MicroRNA Assays in parallel.

Megaplex™ RT Primers are highly multiplexed RT primers designed to convert up to 381 miRNAs to cDNA prior to real-time analysis, and simplify the the TaqMan™ MicroRNA Array v2.0. When sensitivity is of the utmost importance, and/or sample is limiting Megaplex™ PreAmp Primers provide an optional preamplification step. TaqMan™ MicroRNA Arrays v2.0 and Megaplex™ Primer Pools enable a comprehensive expression profile consistent with Sanger miRBase v10 to be created in as little as 5 hours, providing the ideal miRNA profiling solution.

Simplify Sample Handling and Increase Sample Throughput
Following reverse transcription of miRNA targets using Megaplex RT Primers and optional preamplification using Megaplex PreAmp Primers, TaqMan™ Universal PCR Master Mix is simply combined with each reaction and pipetted into each of the eight sample loading ports in the TaqMan™ Array, simplifying sample handling and increasing sample throughput.

Provides Comprehensive Coverage of Sanger miRBase v10
Megaplex™ RT Pools and PreAmp Primers are available for human and rodent species, and consist of pre-selected and matched content across a series of primer pools and TaqMan™ MicroRNA Arrays v2.0. The content is derived from miRBase v10, and provides close to full coverage of Sanger with the most up-to-date TaqMan™ MicroRNA Assays. Each array (either human or rodent) contains a selection of carefully chosen endogenous control assays, providing you with a variety of options for data normalization. Of particular note is the inclusion of a new mammalian U6 assay, repeated four times on each card, and an assay unrelated to mammalian species, ath-miR159a, providing a process control.

Ideal for miRNA Profiling
The highly streamlined workflow—coupled with the high specificity, sensitivity, and dynamic range of TaqMan™ Assays—make this product solution ideal for miRNA profiling, especially when compared to microarrays.
TRUSTED_SUSTAINABILITY

Specifications

Type TaqMan™ Rodent MicroRNA A Array v2.0
Format 384-well Microfluidic Card
Content And Storage 2°C to 8°C
For Use With (Application) Real-time PCR (qPCR), miRNA and non-coding RNA analysis
Includes 384-well arrays and information CD
Quantity 4 Packs

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.